Learn More
Merck MultiScreen™ Sealing Tape
Product Code. 10207820 Shop All Merck Products
Click to view available options
Color:
Clear
Opaque
Unit Size:
Pack of 100
Product Code. 10207820
Brand: Merck MATAHCL00
Choose from opaque or clear
  • Opaque sealing tape is used to seal 100μL plate bottoms read in TopCount™ instruments
  • Clear sealing tape can be used to seal all manufacturers' 300μL plate tops and plate bottoms for MicroBeta™ readers
  • Precut to size
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Sterility Non-sterile
No. per Pack 100
For Use With (Application) Used to seal all manufacturers 300μL plate tops and plate bottoms for MicroBeta™ readers
Product Type Sealing Tape