Learn More
Merck MultiScreen™ 96-Well Assay Plates
Product Code. 11734694 Shop All Merck Products
Change view
Click to view available options
Color:
Clear
White
Product Type:
MultiScreen Acceptor Plate
MultiScreen Plate
MultiScreen Transport Receiver Filter Plate
MultiScreen-IP Filter Plate
MultiScreen-IP Filter Plate w/ Underdrain
Unit Size:
Each
Pack of 10
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type Unit Size
11734694 Clear MultiScreen Plate Pack of 10
10741801 Clear MultiScreen-IP Filter Plate w/ Underdrain Pack of 50
10278471 White MultiScreen Acceptor Plate Each
10164213 Clear MultiScreen-IP Filter Plate Pack of 10
10513224 Clear MultiScreen Transport Receiver Filter Plate Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11734694 Supplier Merck Supplier No. MPC4NTR10
Estimated Shipment: 11-12-2026
Provide tools to perform high-throughput studies of ADME processes—the investigation of the adsorption, distribution, metabolism, and excretion of potential drugs
Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold
Choice of filter materials
  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes
Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.
†Includes lid, filter plate, and receiver plate.
‡Includes extended centrifugal cover, filter plate, and receiver plate.
††Consists of a 96-place holder and a 96-well receiver plate preloaded with Microcon YM-10 filter units.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Pore Size 0.45 μm
Color Clear
Product Type MultiScreen Plate
Material Styrene
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Permeability studies
Quantity 10/Pk.