Description
Provides qualitative measurement of cell monolayer health and quantative measures of confluency. Designed for use with cells cultured in Millicell™ Culture Plate Inserts only.
Utilizes AC current for membrane resistance measurements, producing a zero net charge on cells and eliminating adverse DC current effects on cell membranes. A silver/silver chloride (Ag/AgCI) pellet on each electrode tip measures voltage. Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Warranty
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
Specifications
Specifications
| Product Type | Replacement Electrode |
| For Use With (Equipment) | Millicell-ERS Volt-Ohm Meter |
| For Use With (Application) | To provides qualitative measurement of cell monolayer health and quantative measures of confluency |
| Hertz | 12.5 Hz |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.