Learn More
Merck Immobilon™-P PVDF Membrane
Shop All Merck Products
missing translation for 'orderingAttributeHoverText'
Length (Metric):
10 cm
10 m
15 cm
20 cm
26 cm
26.5 cm
7 cm
8 cm
8.5 cm
9 cm
Width (Metric):
10 cm
12 cm
13.5 cm
15 cm
20 cm
26 cm
3.75 m
8.4 cm
8.5 cm
Format:
Roll
Sheet
missing translation for 'unitSize'
1 roll
Each
Pack of 10
Pack of 50
Description
Immobilon™ PVDF membranes are offered in four types, each optimized for a different protein blotting application. Convenient blotting sandwiches feature pre-cut sheets of membrane and blotting filter paper. Immobilon™ PVDF membranes have high protein adsorption, so you won’t lose proteins during transfer or reprobing. The open pore structure makes it easy to access bound proteins and remove unbound probes. In addition, Immobilon™ PVDF membranes optimized for fluorescent blots dramatically increase signal-to-noise ratios for high sensitivity in quantitative, multiplexing applications.
Specifications
Specifications
| Length (Metric) | 8 cm |
| Width (Metric) | 10 cm |
| For Use With (Application) | Optimized for binding proteins transferred from a variety of gel matricies |
| Format | Sheet |
| Material | PVDF |
| Pore Size | 0.45 μm |