Learn More
Merck Immobilon™-P PVDF Membrane
Product Code. 10663751 Shop All Merck Products
Change view
Click to view available options
Length (Metric):
10 cm
10 m
15 cm
20 cm
26 cm
26.5 cm
7 cm
8 cm
8.5 cm
9 cm
Width (Metric):
10 cm
12 cm
13.5 cm
15 cm
20 cm
26 cm
3.75 m
8.4 cm
8.5 cm
Format:
Roll
Sheet
Unit Size:
1 roll
Each
Pack of 10
Pack of 50
Product Code. Length (Metric) Width (Metric) Format unitSize
10663751 10 cm 10 cm Sheet Pack of 10
10408793 8 cm 10 cm Sheet Pack of 10
10344661 26.5 cm 3.75 m Roll 1 roll
10503224 8.5 cm 13.5 cm Sheet Pack of 10
10505672 7 cm 8.4 cm Sheet Pack of 50
10458712 9 cm 12 cm Sheet Pack of 10
10384511 15 cm 15 cm Sheet Pack of 10
10180771 20 cm 20 cm Sheet Pack of 10
10796851 26 cm 26 cm Sheet Pack of 10
16329601 10 m 8.5 cm Roll Each
10 options
Product Code. 10663751
Brand: Merck IPVH10100
Only null left
Add to basket
Add to basket
Hydrophobic PVDF Transfer Membrane for western blotting
Immobilon™ PVDF membranes are offered in four types, each optimized for a different protein blotting application. Convenient blotting sandwiches feature pre-cut sheets of membrane and blotting filter paper. Immobilon™ PVDF membranes have high protein adsorption, so you won’t lose proteins during transfer or reprobing. The open pore structure makes it easy to access bound proteins and remove unbound probes. In addition, Immobilon™ PVDF membranes optimized for fluorescent blots dramatically increase signal-to-noise ratios for high sensitivity in quantitative, multiplexing applications.
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 10 cm
Width (Metric) 10 cm
For Use With (Application) Optimized for binding proteins transferred from a variety of gel matricies
Format Sheet
Material PVDF
Pore Size 0.45 μm