Learn More
Merck Immobilon™-P Blotting Sandwiches
Product Code. p-3131773 Shop All Merck Products
Change view
Click to view available options
Length (Metric):
7 cm
8.5 cm
Width (Metric):
8.4 cm
13.5 cm
Unit Size:
Pack of 20
Product Code. Length (Metric) Width (Metric) unitSize
10749883 7 cm 8.4 cm Pack of 20
10176172 8.5 cm 13.5 cm Pack of 20
2 options
Product Code. 10749883
Brand: Merck IPSN07852
Immobilon-P membrane interleaved with blotting paper, for standard tank transfer procedures.
Blotting Sandwiches:
  • When you need to process multiple blots, blotting sandwiches are a convenient, time-saving choice. Our sandwiches include sheets of Immobilon-P membrane interleaved with pre-cut sheets of chromatography-grade blotting filter paper and are compatible with any standard wet transfer device.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Length (Metric) 7 cm
    Width (Metric) 8.4 cm
    Material PVDF
    Pore Size 0.45 μm