Learn More
Merck Aerosol Adapter, Luer Slip to 1/4 in.– 3/8 in. ID Hose
Shop All Merck Products
Click to view available options
:
Aerosol Adapter
Description
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications
Specifications
| Connectors | 0.25 to 0.37 in. I.D. Hose |
| For Use With (Equipment) | Millipore Aerosol Analysis Monitor or Swinnex™ 47 mm Membrane Filter Holder |
| Material | Stainless Steel |
| For Use With (Application) | To connect the vacuum source to any millipore aerosol analysis monitor or Swinnex™ 47 mm membrane filter holder |
| Includes | Aerosol Adapter, Luer slip to 1/4 in.- 3/8 in. ID hose, stainless steel |
| Type | Environmental Test Kits or Supplies |
| Diameter (English) Inside | 0.25 in. |
| Diameter (Metric) Inside | 0.63 cm |