Learn More
Merck Accessories for Dispensing Pressure Vessels
Shop All Merck Products
Click to view available options
Type:
Bubble Point Test Apparatus
O-Ring
Pressure Vessel Accessory Kit
Tubing
Unit Size:
Each
Description
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Dispensing Pressure Vessel |
| Sterility | Non-sterile |
| Type | Pressure Vessel Accessory Kit |
| Diameter (English) | 0.25 in. |
| Diameter (Metric) Inside | 6.3 mm |