missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELFN2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15566082
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15566082 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15566082 Supplier Invitrogen™ Supplier No. PA543521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: PVSHPTPYST DAQREPDENS GFNPDEILSV EPPASSTTDA SAGPAIKLHH Sequence homology: Cow: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

ELFN2 is a single-pass membrane protein. It contains 1 fibronectin type-III domain and 5 LRR (leucine-rich) repeats. The exact function of ELFN2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELFN2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ELFN2
Gene Accession No. Q5R3F8
Gene Alias 5leucine rich repeat containing 62; 6330514E13; AW048948; BC094219; dJ63G5.3 (putative Leucine rich protein); Elfn2; Elfn2-ps1; extracellular leucine rich repeat and fibronectin type III domain containing 2; extracellular leucine-rich repeat and fibronectin type III containing 2; extracellular leucine-rich repeat and fibronectin type III domain containing 2; extracellular leucine-rich repeat and fibronectin type III domain containing 2, pseudogene 1; extracellular leucine-rich repeat and fibronectin type III domain-containing protein 2; KIAA1904; leucine rich repeat and fibronectin type III, extracellular 2; leucine rich repeat containing 62; leucine-rich repeat and fibronectin type-III domain-containing protein 6; Leucine-rich repeat-containing protein 62; LOW QUALITY PROTEIN: protein phosphatase 1 regulatory subunit 29; Lrrc62; PPP1R29; protein phosphatase 1 regulatory subunit 29; protein phosphatase 1 regulatory subunit 29-like; protein phosphatase 1, regulatory subunit 29; RGD1559693
Gene Symbols ELFN2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ELFN2 (aa 254-303).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114794
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.