missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55029-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to ELF1.
This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants.Specifications
ELF1 | |
Polyclonal | |
Unconjugated | |
ELF1 | |
E74 like factor 1, ELF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1997 | |
-20°C | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P32519 | |
ELF1 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |