Learn More
Bibby Scientific™ Electrothermal™ Replacement Spare Heater Element
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| For Use With | EMEA3/6/0250 |
| Voltage | 230 V |
| Wattage | 150w |
| Product Type | Replacement Spare Heater Element |
| Frequency | 50 Hz |