Learn More
Cole-Parmer™ Omni Series Reflux/Inert Head
Product Code. 11816742
Change view
Click to view available options
For Use With (Equipment):
For OS1025 STEM Omni Reaction Station, Compatible with Integrity 10, RS-200 and RS-250 Series
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11816742 For OS1025 STEM Omni Reaction Station, Compatible with Integrity 10, RS-200 and RS-250 Series Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11816742 Supplier Cole-Parmer™ Supplier No. 3663072
  • ONLINE EXCLUSIVE PRICE
    26930.00 NOK
    19731.72 NOK / Pack of 1
    Save 7198.28 NOK
    26% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Accessory reflux/inerting head for use with Cole-Parmer™ Integrity 10, RS-200 or RS-250, and OS1025 series reaction stations.
  • Accessory item for reaction stations
  • One year warranty
  • Dimensions (Dia. x L): 10 x 25 mm
  • 10 Positions
Specifications
Product Type Reflux Head
For Use With (Equipment) For OS1025 STEM Omni Reaction Station, Compatible with Integrity 10, RS-200 and RS-250 Series