Learn More
Bibby Scientific™ Electrothermal™ Heating Element for EM/EME
Product Code. 11738412
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
5000 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11738412 1000 mL Each
11748412 2000 mL Each
11768412 5000 mL Each
11876475 250 mL Each
11728412 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11738412 Supplier Bibby Scientific™ Supplier No. 0301255
In Stock
Add to basket
Add to basket
Bibby Scientific Electrothermal Heating Element for EM/EME, Capacity: 250 mL, For Use With: 14314 EM Series Heating Mantles, Voltage: 230 V, Frequency: 50/60 Hz
Specifications
Capacity (Metric) 1000 mL
For Use With EM/EME Series Heating Mantles
Voltage 230 V
Wattage 300w
Product Type Heating Element
Frequency 50/60 Hz