Learn More
3M™ Electronic Test System Unit
Description
- Service parts available to authorized entities only
- Fully automatic with Pass/Fail Bowie-Dick type test result
- Data download to PC software for capture and trending
Specifications
Specifications
| For Use With (Application) | Sterilization Monitoring |
| Model | 4308 |