Learn More
Thermo Scientific™ Electronic Hand-Held Crimpers and Decrimpers, Level 3 High Performance Applications
Product Code. 16366218 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Electronic Crimper
Electronic Decrimper
Power Supply
Size:
11 mm
110/240 V
13 mm
20 mm
8 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Size Unit Size
16366218 Power Supply 110/240 V Each
16397906 Electronic Crimper 11 mm Each
16329236 Electronic Crimper 13 mm Each
16389735 Electronic Crimper 20 mm Each
16339236 Electronic Crimper 8 mm Each
16307916 Electronic Decrimper 11 mm Each
16376218 Electronic Decrimper 13 mm Each
16398145 Electronic Decrimper 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16366218 Supplier Thermo Scientific™ Supplier No. 60180ECRHB
Estimated Shipment: 13-11-2026
Use Thermo Scientific™ Electronic Hand-Held Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Use Thermo Scientific™ Electronic Hand-Held Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively. These hand-held, high-power crimpers and decrimpers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to seal or remove crimp caps with a simple, one-handed push of a button. Use for crimping of vials with aluminum, pressure-release, and bi-metal caps.
The electronic crimper's message window conveniently provides all-important information at one glance. This device is Performance Level 3, which means it is designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses.
Features
Hand-Held Crimper and Decrimper
  • The battery charge icon on the color screen changes to red when battery is low
  • Includes a fully rechargeable lithium ion battery and a universal power supply/recharger with international plug adapters
The Message Window Displays…
  • The actual crimp power
  • An error message after an incorrect crimping process
  • Statistics and log of recent crimping results
  • In the language of your choice (English, Chinese, Japanese, German, French, Spanish, Italian, Korean)
These Electronic Vial Crimpers also feature adjustable settings for best crimp results and a motor with extended lifetime and no particle emission.
TRUSTED_SUSTAINABILITY
Specifications
Power Supply 110/240 V AC
Type Power Supply
For Use With (Equipment) Electronic Crimpers and Decrimpers
Size 110/240 V
Product Line SureSTART Level 3
Quantity Each