Learn More
OHAUS™ Electronic Balance - PR Series
Product Code. 16650304 Shop All Ohaus Products
Change view
Click to view available options
Model:
PR124
PR124/E
PR1602
PR1602/E
PR2202
PR2202/E
PR223
PR223/E
PR224
PR224/E
PR4201
PR4201/E
PR4202
PR4202/E
PR423
PR423/E
PR523
PR523/E
PR6201
PR6201/E
Unit Size:
Each
Product Code. Model unitSize
16650304 PR6201/E Each
16357575 PR224 Each
16381678 PR4202/E Each
16389386 PR4202 Each
16506244 PR2202/E Each
16516244 PR2202 Each
16680294 PR124/E Each
16670294 PR124 Each
16690294 PR224/E Each
16600304 PR223/E Each
16680314 PR223 Each
16610304 PR423/E Each
16690314 PR423 Each
16620304 PR523/E Each
16600324 PR523 Each
16630304 PR1602/E Each
16610324 PR1602 Each
16640304 PR4201/E Each
16620324 PR4201 Each
16630324 PR6201 Each
20 options
Product Code. 16650304
Brand: OHAUS™ 30430078
Designed for Routine Weighing Applications in Your Workplace
  • The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
  • Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
  • The PR's small footprint saves desktop space while providing a large weighing surface. The PR is durably constructed, and features a stainless steel pan to withstand day-to-day use in the workplace.
TRUSTED_SUSTAINABILITY
Specifications
Connection Type RS232 (one port included)
Description User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator
Capacity 6200.00 g
Display Type Backlit LCD
Readability 0.1000 g
For Use With (Application) Basic Weighing, Parts Counting, Percent Weighing
Linearity 0.2000 g
Model PR6201/E
Repeatability 0.1000 g
Stabilization Time 1.000 s
Weighing Modes Milligram, Gram, Kilogram, Ounce, Pound, Carat, Pennyweight, Ounce Troy, Newton, & Grain
Dimensions (L x W x H) 55.000 x 39.000 x 29.000 cm
Length (English) 12.50 in.
Length (Metric) 317.00 mm
Width (English) 7.90 in.
Width (Metric) 201.00 mm
Height (English) 3.70 in.
Height (Metric) 93.000 mm
Depth (English) 12.50 in.
Depth (Metric) 317.00 mm
Weight (English) 7.700 lb.
Weight (Metric) 3.500 kg
Electrical Requirements AC Adapter (included)