Learn More
Eldecalcitol-d6, MedChemExpress
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Eldecalcitol-d6 is the deuterium labeled Eldecalcitol. Eldecalcitol is an orally active analogue of active vitamin D used in the treatment of osteoporosis.
Specifications
Specifications
| Chemical Name or Material | Eldecalcitol-d6 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C30H44D6O5 |
| Color | White |
| Solubility Information | DMSO : 25 mg/mL (50.33 mM; Need ultrasonic) |
| SMILES | C=C1[C@@H](O)[C@H](OCCCO)[C@H](O)C/C1=C/C=C2CCC[C@@]3(C)[C@@]\2([H])CC[C@@H]3[C@@H](CCCC(O)(C([2H])([2H])[2H])C([2H])([2H])[2H])C |
| Industry Type | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 496.75 |
| Quantity | 1 mg |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.