Learn More
Kolmi Hopen™ Elasticated SMS Coveralls with Hood
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Hooded model with flap on zipper
- SMS material makes it possible to better limit release of particles (lint-free)
- Internal seams
- Elasticated with thumb loops at wrists and ankles
- Without pockets to avoid risk of falling objects
- Air and water vapor permeable
- Good tear and abrasion resistance
- Individual packaging
Specifications
Product Type | Cleanroom Coveralls |
Size | 2X-Large |
Material | SMS |
Color | Azure Blue |
Disposable | Single-use |
Hood Type | Elastic |
Sterility | Non-sterile |
Certifications/Compliance | ISO 9001, (EU) 2016/425 |
For Use With (Application) | Controlled Environments |
Weight (Metric) | 50 g/m2 |