missing translation for 'onlineSavingsMsg'
Learn More

ELAC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26639-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895045

  • 3828.53 NOK / 150µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Zinc phosphodiesterase, which displays some tRNA 3′-processing endonuclease activity. Probably involved in tRNA maturation, by removing a 3′-trailer from precursor tRNA.
Specifications

Specifications

ELAC1
Polyclonal
Unconjugated
ELAC1
D29, Deleted in Ma29, elaC homolog 1 (E. coli), ElaC homolog protein 1, ELAC1, Ribonuclease Z 1, RNase Z 1, tRNA 3 endonuclease 1, tRNase Z 1
Rabbit
Antigen Affinity Chromatography
RUO
114615, 55520
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8VEB6, Q9H777
ELAC1
ELAC1 Fusion Protein Ag23939
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers