missing translation for 'onlineSavingsMsg'
Learn More

ELAC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16805055 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16805055 20 μL 20µL
16895045 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16805055

Brand: Proteintech 266391AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Zinc phosphodiesterase, which displays some tRNA 3′-processing endonuclease activity. Probably involved in tRNA maturation, by removing a 3′-trailer from precursor tRNA.

Specifications

Antigen ELAC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ELAC1
Gene Accession No. Q8VEB6, Q9H777
Gene Alias D29, Deleted in Ma29, elaC homolog 1 (E. coli), ElaC homolog protein 1, ELAC1, Ribonuclease Z 1, RNase Z 1, tRNA 3 endonuclease 1, tRNase Z 1
Gene Symbols ELAC1
Host Species Rabbit
Immunogen ELAC1 Fusion Protein Ag23939
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114615, 55520
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.