missing translation for 'onlineSavingsMsg'
Learn More

EIPA, MedChemExpress

Product Code. p-200100282
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16486327

Brand: Medchem Express HY101840

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EIPA (L593754) is an orally active TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA also enhances autophagy by inhibiting Na+/H+-exchanger 3 (NHE3). EIPA inhibits macropinocytosis as well. EIPA can be used in the research of inflammation and cancers, such as gastric cancer, colon carcinoma, pancreatic carcinoma.

EIPA (L593754) is an orally active TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA also enhances autophagy by inhibiting Na+/H+-exchanger 3 (NHE3). EIPA inhibits macropinocytosis as well. EIPA can be used in the research of inflammation and cancers, such as gastric cancer, colon carcinoma, pancreatic carcinoma.

Specifications

Chemical Name or Material EIPA
CAS 1154-25-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H18ClN7O
Quantity 5 mg
Synonym L593754 MH 12-43
Solubility Information DMSO : 140 mg/mL (467.04 mM; Need ultrasonic)
SMILES O=C(C1=NC(Cl)=C(N(CC)C(C)C)N=C1N)NC(N)=N
Molecular Weight (g/mol) 299.76
Formula Weight 299.76
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.