missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | EIF4B |
|---|---|
| Concentration | 0.45 mg/mL |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857924
|
Proteintech
17917-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847924
|
Proteintech
17917-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF4B is a multidomain eukaryotic translation initiation factor with a range of activities. It contains one RRM (RNA recognition) domain, a consensus RNA-binding site (RNP-CS) near the N terminus, and a central domain rich in aspartic acid, arginine, tyrosine, and glycine (DRYG). The DRYG domain of eIF4B helps in self-association and interaction with the p170 subunit of eIF3. eIF4B binds to both mRNA and 18S ribosomal RNA and thus acts as a bridge between the mRNA and the 40S ribosomal subunit during translation initiation. The ATPase and RNA helicase activity of the DEAD box protein eIF4A is stimulated by eIF4B and thus functions in close association with eIF4F a heterotrimeric protein that binds to mRNA cap structure and unwinds inhibitory secondary structure in the 5′ untranslated region of eucaryotic mRNAs in an ATP-dependent manner. Ubiquitous expression is seen in most of the tissues.Specifications
| EIF4B | |
| Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P23588, Q8BGD9 | |
| 1975, 300253, 75705 | |
| EIF4B Fusion Protein Ag12325 | |
| Primary | |
| -20°C | |
| EIF4B |
| 0.45 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| EIF 4B, EIF4B, PRO1843 | |
| Eif4b | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title