missing translation for 'onlineSavingsMsg'
Learn More

EIF3A Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30498212 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498212 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498212 Supplier Novus Biologicals Supplier No. NBP335076AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EIF3A Polyclonal antibody specifically detects EIF3A in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF3A
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias centrosomin homolog, cytoplasmic protein p167, eIF3 p167, eIF3 p180, eIF3 p185, EIF3, p180 subunit, eIF3a, eIF3-p170, EIF3S10EIF3, eIF-3-theta, eIF3-theta, Eukaryotic translation initiation factor 3 subunit 10, eukaryotic translation initiation factor 3 subunit A, eukaryotic translation initiation factor 3, subunit 10 (theta, 150/170kD), eukaryotic translation initiation factor 3, subunit 10 (theta, 170kD), eukaryotic translation initiation factor 3, subunit 10 theta, 150/170kDa, eukaryotic translation initiation factor 3, subunit 10, 170kD, eukaryotic translation initiation factor 3, subunit A, KIAA0139P167, p180, p185, TIF32
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human EIF3A (NP_003741.1).,, Sequence:, MPAYFQRPENALKRANEFLEVGKKQPALDVLYDVMKSKKHRTWQKIHEPIMLKYLELCVDLRKSHLAKEGLYQYKNICQQVNIKSLEDVVRAYLKMAEEKTEAAKEESQQMVLDIEDLDNIQTPESVLLSAVSGEDTQDRTDRLLLTPWVKFLWESYRQCLDLLRNNSRVERLYHDIAQQAFKFCLQYTRKAEFRKLCDNLRMHLSQIQRHHNQSTAINLNNPESQSMHLETRLVQLDSAISMELWQEAF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8661
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.