missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2C1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19690-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to EIF2C1.
AGO1 is a member of the Argonaute family of proteins which play a role in RNA interference. The protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing.Specifications
| EIF2C1 | |
| Polyclonal | |
| Unconjugated | |
| AGO1 | |
| AGO1, Argonaute1, eIF 2C 1, EIF2C, eIF2C 1, EIF2C1, GERP95, hAgo1, Protein argonaute 1, Q99 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 236511, 26523, 313594 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CJG1, Q9UL18 | |
| Ago1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction