missing translation for 'onlineSavingsMsg'
Learn More

EIF2C1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16849164 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16849164 150 μL 150µL
16859164 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16849164

Brand: Proteintech 196901AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody is specific to EIF2C1.

AGO1 is a member of the Argonaute family of proteins which play a role in RNA interference. The protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing.

Specifications

Antigen EIF2C1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AGO1
Gene Accession No. Q8CJG1, Q9UL18
Gene Alias AGO1, Argonaute1, eIF 2C 1, EIF2C, eIF2C 1, EIF2C1, GERP95, hAgo1, Protein argonaute 1, Q99
Gene Symbols Ago1
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 236511, 26523, 313594
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.