missing translation for 'onlineSavingsMsg'
Learn More

eIF2B epsilon Antibody [Alexa Fluor« 700], Novus Biologicals Biologicals™

Product Code. 30499590 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499590 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499590 Supplier Novus Biologicals Supplier No. NBP335116AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

eIF2B epsilon Polyclonal antibody specifically detects eIF2B epsilon in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF2B epsilon
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CACH, CLE, EIF-2B, eIF-2B GDP-GTP exchange factor subunit epsilon, EIF2BE, EIF2Bepsilon, eukaryotic translation initiation factor 2B, subunit 5 (epsilon, 82kD), eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa, LVWM, translation initiation factor eIF-2B subunit epsilon
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 442-721 of human eIF2B epsilon (eIF2B epsilon (EIF2B5)) (NP_003898.2).,, Sequence:, TLPEGSVISLHPPDAEEDEDDGEFSDDSGADQEKDKVKMKGYNPAEVGAAGKGYLWKAAGMNMEEEEELQQNLWGLKINMEEESESESEQSMDSEEPDSRGGSPQMDDIKVFQNEVLGTLQRGKEENISCDNLVLEINSLKYAYNISLKEVMQVLSHVVLEFPLQQMDSPLDSSRYCALLLPLLKAWSPVFRNYIKRAADHLEALAAIEDFFLEHEALGISMAKVLMAFYQLEILAEETILSWFSQRDTTDKGQQLRKNQQLQRFIQWLKEAEEESSEDD
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8893
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.