Learn More
EGF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Description
AA Sequence of recombinant protein: MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat EGF. Anti-Rat EGF-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat EGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat EGF (500-P277) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat EGF. Western Blot: To detect Rat EGF by Western Blot analysis this antibody can be used at a concentration of 0.5-1.0 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat EGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P277BT-1MG will be provided as 2 x 500 μg
Specifications
Specifications
| Antigen | EGF |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with no preservative |
| Gene | EGF |
| Gene Accession No. | P07522 |
| Gene Alias | AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone |
| Gene Symbols | EGF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.