Learn More
EFHD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16889733 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16889733 150 μL 150µL
16899733 20 μL 20µL
2 options
Product Code. 16889733
Brand: Proteintech 121811AP150UL
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rabbit Polyclonal Antibody
EFHD2, also known as Swiprosin-1 or SWS1, is an EF-hand and coiled-coil-containing adaptor protein that plays a role in lymphocyte physiology. EFHD2 exhibits the highest expression in CD8+ T cells and immature B cells. It provides a membrane scaffold that is required for the Syk-, SLP-65-, and PLC-gamma2-dependent B-cell receptor (BCR)-induced calcium flux. EFHD2 may also regulate BCR-induced immature and primary B-cell apoptosis. It controls spontaneous apoptosis through the regulation of BCL2L1 abundance. Also, EFHD2 plays a role as negative regulator of the canonical NF-kappa-B-activating branch.
Specifications
Antigen EFHD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EFHD2
Gene Accession No. Q96C19, Q9D8Y0
Gene Alias EFHD2, Swiprosin 1, SWS1
Gene Symbols Efhd2
Host Species Rabbit
Immunogen EFHD2 Fusion Protein Ag2826
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27984, 79180
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG