missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCAB2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25156-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
EFCAB2 | |
Polyclonal | |
Unconjugated | |
EFCAB2 | |
EFCAB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84288 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VUJ9 | |
EFCAB2 | |
EFCAB2 Fusion Protein Ag18372 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |