missing translation for 'onlineSavingsMsg'
Learn More

EEPD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 24310-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892785

  • 4593.76 NOK / 150µL
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

EEPD1 gene ontology annotations related to this gene include DNA repair.
Specifications

Specifications

EEPD1
Polyclonal
Unconjugated
EEPD1
EEPD1, HSPC107, KIAA1706
Rabbit
Antigen Affinity Chromatography
RUO
67484, 80820
-20°C
Liquid
Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3TGW2, Q7L9B9
Eepd1
EEPD1 Fusion Protein Ag19423
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers