missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEPD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24310-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
EEPD1 gene ontology annotations related to this gene include DNA repair.Specifications
EEPD1 | |
Polyclonal | |
Unconjugated | |
EEPD1 | |
EEPD1, HSPC107, KIAA1706 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67484, 80820 | |
-20°C | |
Liquid |
Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3TGW2, Q7L9B9 | |
Eepd1 | |
EEPD1 Fusion Protein Ag19423 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |