missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10630-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The calculated molecular weight of EEF1D is 31 kDa, but the modified protein is about 35-40 kDa.
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.Specifications
EEF1D | |
Polyclonal | |
Unconjugated | |
EEF1D | |
Antigen NY CO 4, EEF1D, EF 1 delta, EF 1D, EF1D, Elongation factor 1 delta, FP1047 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1936, 300033, 66656 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29692, P57776, Q68FR9 | |
Eef1d | |
EEF1D Fusion Protein Ag0983 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |