missing translation for 'onlineSavingsMsg'
Learn More

EEF1D Mouse anti-Human, Mouse, Rat, Clone: 3G4C4, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60085-2-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887055

  • 3468.15 NOK / 150µL
Estimated Shipment: 13-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The calculated molecular weight of EEF1D is 31 kDa, but the modified protein is about 35-40 kDa.

This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit, delta, functions as guanine nucleotide exchange factor. It is reported that following HIV-1 infection, this subunit interacts with HIV-1 Tat. This interaction results in repression of translation of host cell proteins and enhanced translation of viral proteins. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene. Related pseudogenes have been defined on chromosomes 1, 6, 7, 9, 11, 13, 17, 19.
Specifications

Specifications

EEF1D
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P29692, P57776, Q68FR9
Eef1d
EEF1D Fusion Protein Ag0983
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG1
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
3G4C4
Unconjugated
EEF1D
Antigen NY CO 4, EEF1D, EF 1 delta, EF 1D, EF1D, Elongation factor 1 delta, FP1047
Mouse
Antigen Affinity Chromatography
RUO
1936, 300033, 66656
-20°C
Liquid
Videos
SDS
Documents
Certificates
Special Offers

Special Offers