Learn More
EDTA Disodium Salt Solution, 0.05M, Chem-Lab
Product Code. 10778654
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10778654 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10778654 Supplier Chem-Lab Supplier No. CL05.0505.1000
  • ONLINE EXCLUSIVE PRICE
    312.00 NOK
    253.60 NOK / 1 L
    Save 58.40 NOK
    18% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Used in electrophoresis and molecular biology applications. EDTA Disodium Salt Solution, 0.05M, Chem-Lab is a chelator of divalent cations for use in TH titrations. Solution contains 18.612g EDTA diNa · 2H2O per liter of water (0.01 M).
EDTA Disodium Salt Dihydrate is a chelator of divalent cations. It inhibits enzymes, such as metalloproteases, that require divalent cations for activity. It is also used in many enzyme buffers and as an enzyme inactivator at high concentrations.
Specifications
CAS 6381-92-6
Melting Point 0°C
Color Colorless
Density 1.01g/cm3
pH 4 to 5
Boiling Point 100°C
Odor Odorless
Molecular Formula C10H18N2Na2O10
MDL Number MFCD00150037,MFCD00003541
Quantity 1 L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
Solubility Information Soluble
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Molecular Weight (g/mol) 372.24
PubChem CID 44120005
Formula Weight 372.24g/mol
Concentration or Composition (by Analyte or Components) 0.05mol/L (± 0.0004, 20°C)
Physical Form Liquid
Show More Show Less
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.