Learn More
Velp Scientifica™ EDTA-Certified Calibration Standard
Product Code. 15319574 Shop All Velp Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15319574 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15319574 Supplier Velp Scientifica™ Supplier No. A00000149
  • ONLINE EXCLUSIVE PRICE
    1653.30 NOK
    1555.94 NOK / 100g
    Save 97.36 NOK
    5% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Velp Scientifica™ Carbon, Hydrogen, and Nitrogen standard to create and check calibration curves of Elemental Analyzers.
  • For use with NDA 701 and 702
Specifications
Type Calibration Standard
For Use With Dumas Nitrogen Analyzer
Quantity 100 g
Certifications/Compliance EDTA Certified
compliance-icons compliance-icons
Product Identifier
  • EDTA, 100 G
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: EDTA