Learn More
Edoxaban tosylate, MedChemExpress
Product Code. 16466477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466477 5 mg 5mg
16464816 100 mg 100mg
16440137 50 mg 50mg
16403758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16466477 Supplier Medchem Express Supplier No. HY10264A
MedChemExpress Edoxaban (DU-176b) tosylate is an orally active, highly potent, selective, and direct Factor Xa (FXa) inhibitor with Kis of 0.561 and 2.98 nM for free human FXa and prothrombinase. Edoxaban tosylate exhibits more than 10,000-fold selectivity over other coagulation proteases. Edoxaban tosylate can be used for preventing thromboembolic disease research.
Edoxaban (DU-176b) tosylate is an orally active, highly potent, selective, and direct Factor Xa (FXa) inhibitor with Kis of 0.561 and 2.98 nM for free human FXa and prothrombinase. Edoxaban tosylate exhibits more than 10,000-fold selectivity over other coagulation proteases. Edoxaban tosylate can be used for preventing thromboembolic disease research.
Specifications
Chemical Name or Material Edoxaban tosylate
CAS 480449-71-6
Color White
Purity Grade Notes Research
Molecular Formula C31H38ClN7O7S2
Quantity 5 mg
Synonym DU-176b
Solubility Information DMSO : 100 mg/mL (138.84 mM; Need ultrasonic)
SMILES O=C(N(C)C)[C@@H](CC1)C[C@H]([C@H]1NC(C(NC(C=C2)=NC=C2Cl)=O)=O)NC(C3=NC(CC4)=C(CN4C)S3)=O.O=S(C5=CC=C(C)C=C5)(O)=O
Molecular Weight (g/mol) 720.26
Formula Weight 720.26
Percent Purity 99.47%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation