Learn More
Ansell EDGE™ 48-707 Cut and Abrasion-resistant Gloves
Product Code. 18866155 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
Unit Size:
144 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18866155 10 144 pairs
18826155 6 144 pairs
18836155 7 144 pairs
18846155 8 144 pairs
18856155 9 144 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 18866155 Supplier Ansell Supplier No. 4870710
  • ONLINE EXCLUSIVE PRICE
    18645.15 NOK
    12103.00 NOK / 144 pairs
    Save 6542.15 NOK
    35% Off
Estimated Shipment: 04-12-2026
Affordable safety gloves, offering superb grip as well as cut and abrasion resistance.
  • Advanced protection: EDGE™ 48-707 cut and abrasion-resistant gloves provide ANSI cut level A6 and EN 388 ISO cut level F, which afford a very high level of cut protection
  • Heightened grip and dexterity: Owing to their palm PU coating, these protective gloves combine good grip as well as dexterity, and are suitable for dry or slightly oily environments
  • Reduced risks: In addition, EDGE™ 48-707 gloves meet electrostatic discharge protection needs and feature anti-static capabilities.
  • Added features: These grey hand protection gloves also possess a seamless liner and knitted wrist, which deliver a secure fit for use in applications that require high cut resistance
TRUSTED_SUSTAINABILITY
Specifications
Coating Material Polyurethane
Color Black
Cut Resistance Rating Level F
Hand Specific
Latex Free Yes
Material Polyester, Fiberglass, Spandex, Nylon, HPPE
Product Type Industrial Gloves
Size 10