Learn More
Edetate Calcium Disodium Dihydrate, TRC
Product Code. 30132492
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132492 10 g 10g
30136235 50 g 50g
30006092 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132492 Supplier Toronto Research Chemicals Supplier No. TRCE55559310G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6766-87-6
Molecular Formula C10H12CaN2O8 . 2H2O . 2Na
Molecular Weight (g/mol) 410.3
Synonym Edetate Calcium Disodium(Secondary Standards Traceble To Usp)
SMILES O.O.[Na+].[Na+].[Ca+2].[O-]C(=O)CN(CCN(CC(=O)[O-])CC(=O)[O-])CC(=O)[O-]
Specifications
Quantity 10 g
InChI Formula InChI=1S/C10H16N2O8.Ca.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;2*1H2/q;+2;2*+1;;/p-4
Formula Weight 410.0226
Chemical Name or Material Edetate Calcium Disodium Dihydrate