Learn More
ECOSURF™ EH-9 Surfactant, Thermo Scientific™
Product Code. p-7222014
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
Product Code. Quantity unitSize
17207056 100 mL 100mL
17217056 500 mL 500mL
17297046 2500 mL 2500mL
3 options
Product Code. 17207056
Brand: Thermo Scientific Alfa Aesar H37158.AE
Request Bulk or Custom Format
ECOSURF™ EH-9 Surfactants are included in a group of chemicals that align with the 10th principle of green chemistry.
Range of surfactants that are included in a group of chemicals that align with the 10th principle of green chemistry: Design for Degradation.
  • These chemical products are designed to break down to innocuous substances after use to avoid accumulation and preventing harm to the environment.
  • ECOSURF™ EH-9 Surfactant is one such alternative detergent to octylphenol ethoxylate containing detergents, like e.g. Triton X™-100.
  • This is a biodegradable surfactant to reduce your ecological footprint.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64366-70-7
Molecular Formula C13H28O3
Molecular Weight (g/mol) 232.36
InChI Key LLCXFSRLSYMSPW-UHFFFAOYNA-N
IUPAC Name 2-ethylhexan-1-ol; 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1.CCCCC(CC)CO
Specifications
Color Clear colorless to pale yellow
Physical Form Liquid
Cloud Point 10% in water: 62 to 67°C
Density 1.026 g/mL
pH 5.0 to 7.5 (1% aq. soln.)
Quantity 100 mL
Flash Point 288°C (550°F)