Learn More
Norell™ Economy Standard Borosilicate Glass NMR Sample Tubes
Product Code. 12872336
Change view
Click to view available options
Length (Metric):
178 mm
203 mm
Diameter (Metric) Outer:
4.97 mm
10 mm
Packaging:
100 Pack
5 Pack
50 Pack
Unit Size:
Pack of 100
Pack of 5
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Outer Packaging Unit Size
12872336 203 mm 4.97 mm 5 Pack Pack of 5
12820026 178 mm 10 mm 100 Pack Pack of 100
12830026 203 mm 10 mm 100 Pack Pack of 100
12852336 203 mm 4.97 mm 50 Pack Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12872336 Supplier Norell™ Supplier No. 5528
  • ONLINE EXCLUSIVE PRICE
    341.00 NOK
    297.05 NOK / Pack of 5
    Save 43.95 NOK
    12% Off
View all promotions
Estimated Shipment: 22-12-2026
For auto-sampling systems and high-throughput NMR
  • For auto-sampling systems and high-throughput NMR
  • The Norell Standard Series™ of NMR tubes has been developed for quality and precision
  • Virtually all bottom and wall variation has been eliminated, greatly improving shimmering times and sample reproducibility
  • Whether monitoring a reaction, scouting for a new compound, or doing routine NMR, the Standard Series™ is the tube of choice for today's scientist
  • All tubes have polished ends making them less susceptible to breaking
  • Colored caps are included
Specifications
Concentricity (Metric) 0.070 mm
Length (Metric) 203 mm
Material Borosilicate Glass
Product Type NMR Sample Tube
Packaging 5 Pack
Diameter (Metric) Outer 4.97 mm
Diameter (Metric) Inner 4.2 mm
Includes Caps