Learn More
Cole Parmer™ Economical Polypropylene Graduated Cylinder
Product Code. p-7161079 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
10 mL
50 mL
100 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
15777968 10 mL Each
15787968 - Each
15797968 50 mL Each
15707978 100 mL Each
4 options
Product Code. 15777968
Brand: Cole Parmer™ 0613770
Only null left
Add to basket
Add to basket
Polypropylene (PP) cylinders offer very good chemical resistance.
  • Meet ISO Standard 6706.
  • Calibrated to contain for measuring final volumes.
  • Molded graduations for durability.
  • Polypropylene construction provides very good chemical resistance.
Specifications
Capacity (Metric) 10 mL
Material Polypropylene
Certifications/Compliance ISO 6706
Calibration To Contain
Autoclavable Autoclavable
Shape Round
Base Type Hexagonal
Type Graduated Cylinder