Learn More
Ansell™ Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves
Product Code. p-8005174 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
Product Code. Size unitSize
12697011 10 12 pairs
12667011 7 12 pairs
12677011 8 12 pairs
12687011 9 12 pairs
4 options
Product Code. 12697011
Brand: Ansell™ 87190100BP
Only null left
Add to basket
Add to basket
Econohands™ 87-190 Series Yellow Natural Rubber Latex Extra Thin Gloves - Extra thin glove for excellent sensitivity. Specially treated to reduce the risk of allergic reactions.
  • Specially treated to reduce the risk of allergic reactions.
  • Thickness of 0.35 mm, provides outstanding sensitivity.
  • Excellent price/quality ratio.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Rubber Latex
Cuff Material Natural Rubber Latex
Applications Industrial
Coating Material Natural Rubber Latex
Color Yellow
Latex Free No
Material Rubber Latex
Palm Thickness (English) 0.0138 in.
Palm Thickness (Metric) 0.35 mm
Product Type Extra Thin Gloves
Size 10
Thickness (English) 0.0138 in.
Thickness (Metric) 0.35 mm
Glove Cuff Color Yellow
Glove Cuff Style Straight
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Liquid & mechanical resistant
Series 87-190