Learn More
Thermo Scientific™ Eco32I (EcoRV) (10 U/μL)
Product Code. p-4521757 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2000 U
4,000 units
5 x 2,000 units
Unit Size:
2000 units
4000 units
Pack of 5
Product Code. Quantity unitSize
10369529 2000 U 2000 units
11873823 5 x 2,000 units Pack of 5
10829180 4,000 units 4000 units
3 options
Product Code. 10369529
Brand: Thermo Scientific™ ER0301
Edge
The Eco32I (EcoRV) restriction enzyme recognizes GAT^ATC sites and cuts best at 37°C in R buffer (Isoschizomers: EcoRV).
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Applications
  • Molecular cloning
  • Restriction site mapping
  • Genotyping
  • Southern Blot
  • Restriction fragment length polymorphism (RFLP)
  • SNP
TRUSTED_SUSTAINABILITY
Specifications
Concentration 10 U/μL
Incubator Temperature 37°C
Quantity 2000 U
Product Type Eco32I (EcoRV)