Learn More
Fisherbrand™ ECO 25 Thermoreactor for COD Analysis
Product Code. 11751304 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Range:
70°, 100°, 120°, 150°, 160°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
11751304 70°, 100°, 120°, 150°, 160°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11751304
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Enables the user to determine parameters such as COD, nitrogenous and phosphorated compounds. Fisherbrand™ ECO 25 Thermoreactor for COD Analysis enable the preparing of samples by way of wet digestion for determining metallic and non-metallic elements in organic and inorganic materials.
  • COD analyzed in just 30 minutes
  • Acoustic signal and automatic switch-off at the end of each cycle
  • Five working selectable temperatures: (70°, 100°, 120°, 150° and 160°C)
  • Test tube cover, heating block protection and safety thermostat to enhance safety and reliability
Specifications
Description Thermoreactor
Range 70°, 100°, 120°, 150°, 160°C