Learn More
Tenak™ Eco AluRack for Upright Freezers
Product Code. 12422570 Shop All TENAK Products
Click to view available options
Holds:
Boxes (12)
Boxes (15)
Boxes (8)
Dimensions (L x W x H):
558 x 139 x 264 mm
558 x 139 x 268 mm
688 x 139 x 240 mm
Unit Size:
Each
Product Code. 12422570
Brand: Tenak™ TE11344
Only null left
Add to basket
Add to basket
Providing a whole new way of making storage racks. Tenak™ Eco AluRacks for Upright Freezers fit into any freezer and can be used with all standard cryoboxes.
  • Made of seawater resistant aluminum
  • Available in a range of box heights
  • Racks can be used with all freezer brands
  • Sold without cryoboxes
Specifications
Product Type Eco AluRack
Description For boxes 130mm high
Array 3 x 3
Holds Boxes (8)
Material Aluminum
Shelves 9
Stackable Yes
For Use With (Equipment) For upright freezers
No. per Pack 1 Pack
Quantity 1 Pack
Dimensions (L x W x H) 558 x 139 x 268 mm
Width (Metric) 139 mm
Height (Metric) 268 mm
Depth (Metric) 558 mm