Learn More
Tenak™ ECO AluRack for Upright Freezers, 130 mm Box Height, with Boxes
Shop All TENAK Products
Click to view available options
Holds:
Boxes
Unit Size:
Each
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Specifications
Specifications
| Includes | White cardboard boxes, 9 x 9 cell dividers |
| Description | Seawater resistant |
| Array | 3 x 5 |
| Holds | Boxes |
| Material | Aluminum |
| Positions | 15 |
| For Use With (Equipment) | Upright Freezers |
| Dimensions (L x W x H) | 683 x 139 x 401 mm |
| Height (Metric) | 130 mm (Box) |