Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Specifications
Specifications
| Description | Seawater resistant |
| Holds | Boxes |
| Material | Aluminum |
| Positions | 6 |
| For Use With (Equipment) | Upright Freezers |
| Dimensions (L x W x H) | 425 x 139 x 268 mm |
| Includes | White cardboard boxes, 9 x 9 cell dividers |
| Height (Metric) | 130 mm (Box) |
| Array | 2 x 3 |