missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 25 mm Shelf Height

Eco AluRack for upright freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE15470

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12468230

  • 2140.00 NOK / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications

Specifications

25 mm
Upright Freezers
15 x 5
450 x 135 x 398 mm
Microtiterplates (75)
Aluminum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers