Learn More
Tenak™ Eco AluRack for Microtiter Plates for Upright Freezers
Providing a complete range of racks for microtiter plates in upright freezers. Tenak™ Eco AluRacks for Microtiter Plates for Upright Freezers are stackable for convenience.
Brand: Tenak™ TE15452
Additional Details : Weight : 1.60000kg
Description
- A range of sizes available
- Racks can either be used as one plate per shelf or the plates can be stacked
- Racks can be used with all freezer models
- Manufactured from aluminum
Specifications
Microtiter Plate Rack | |
13 x 5 | |
Aluminum | |
Microtiter Plates, Upright Freezers | |
450 x 135 x 221mm | |
221 mm | |
450 mm |
For shelves 16mm high | |
Plates (65) | |
Yes | |
1 Pack | |
135 mm | |
16 mm |