Learn More
Sharpsguard™ Eco 11.5 Waste Container
Product Code. 17754854
Change view
Click to view available options
Color:
Orange
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17754854 Orange Each
17764854 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17754854 Supplier Sharpsguard™ Supplier No. DD476OLR
  • ONLINE EXCLUSIVE PRICE
    68.16 NOK
    62.11 NOK / Pack of 1
    Save 6.05 NOK
    8% Off
Estimated Shipment: 29-12-2026
The new Sharpsguard™ eco containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
  • Sharpsguard™ eco 11.5 is a multi-purpose container featuring a robust pail handle for easy transportation. A wider aperture allows easy insertion of larger items.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-0
Specifications
Capacity (Metric) 9.27 L
Diameter (Metric) Outer 275 mm
Color Orange
Height (Metric) 249 mm
Quantity 1 Ea.
Weight (Metric) 622 g